5-Amino-2-methylbenzotrifluorid

Lieferant: FLUOROCHEM

Synonyme: 4-Methyl-3-(trifluoromethyl)aniline

010687-5G
FLUO010687-5GEA 97.1 EUR
FLUO010687-5G
5-Amino-2-methylbenzotrifluorid
5-Amino-2-methylbenzotrifluorid
Formel: C₈H₈F₃N
Molecular Weight: 175,16 g/mol
MDL: MFCD01631582
CAS-Nummer: 65934-74-9
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR