6-Aminonicotinsäure

Lieferant: FLUOROCHEM

Synonyme: 6-Amino-3-pyridincarbonsäure, 6-Aminopyridin-3-carbonsäure

010692-5G 010692-25G
FLUO010692-5GEA 0 EUR
FLUO010692-5G FLUO010692-25G
6-Aminonicotinsäure
6-Aminonicotinsäure
Formel: C₆H₆N₂O₂
Molecular Weight: 138.13 g/mol
Schmelzpunkt: > 300 °C
MDL: MFCD00006326
CAS-Nummer: 3167-49-5
EINECS: 221-630-4
Merck Index: 13,00455

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR