2-Chlor-4-fluor-5-nitrophenol

Lieferant: FLUOROCHEM

010722-1G
FLUO010722-1GEA 31.1 EUR
FLUO010722-1G
2-Chlor-4-fluor-5-nitrophenol
2-Chlor-4-fluor-5-nitrophenol
Formel: C₆H₃ClFNO₃
Molecular Weight: 191,5400 g/mol
MDL: MFCD02670258
CAS-Nummer: 84478-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR