2-Brom-4-(trifluormethyl)anisol

Lieferant: FLUOROCHEM

010726-5G 010726-25G
FLUO010726-5GEA 73.5 EUR
FLUO010726-5G FLUO010726-25G
2-Brom-4-(trifluormethyl)anisol
2-Brom-4-(trifluormethyl)anisol
Formel: C₈H₆BrF₃O
MDL: MFCD03789249
CAS-Nummer: 402-10-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR