3-Fluor-4-(trifluormethyl)pyridin
Lieferant: FLUOROCHEM
010727-1G
010727-2.5G
FLUO010727-1GEA
60.9
EUR
FLUO010727-1G
FLUO010727-2.5G
3-Fluor-4-(trifluormethyl)pyridin
3-Fluoro-4-(trifluoromethyl)pyridine
Formel:
C₆H₃F₄N Molecular Weight: 165.09 g/mol |
MDL:
MFCD03789250 CAS-Nummer: 113770-87-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...