3-Fluor-4-(trifluormethyl)pyridin

Lieferant: FLUOROCHEM

010727-1G 010727-2.5G
FLUO010727-1GEA 60.9 EUR
FLUO010727-1G FLUO010727-2.5G
3-Fluor-4-(trifluormethyl)pyridin
3-Fluoro-4-(trifluoromethyl)pyridine
Formel: C₆H₃F₄N
Molecular Weight: 165.09 g/mol
MDL: MFCD03789250
CAS-Nummer: 113770-87-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR