4-Fluor-2-benzo[b]thiophencarbonsäure

Lieferant: FLUOROCHEM

010734-1G
FLUO010734-1GEA 213 EUR
FLUO010734-1G
4-Fluor-2-benzo[b]thiophencarbonsäure
4-Fluor-2-benzo[b]thiophencarbonsäure
Formel: C₉H₅FO₂S
Molecular Weight: 196.1982 g/mol
MDL: MFCD03425782
CAS-Nummer: 310466-37-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR