2-Chlor-4-fluor-5-nitroanisol

Lieferant: FLUOROCHEM

Synonyme: 1-Chloro-5-fluoro-2-methoxy-4-nitrobenzene

010744-1G
FLUO010744-1GEA 67.1 EUR
FLUO010744-1G
2-Chlor-4-fluor-5-nitroanisol
2-Chlor-4-fluor-5-nitroanisol
Formel: C₇H₅ClFNO₃
Molecular Weight: 205,5700 g/mol
MDL: MFCD03425810
CAS-Nummer: 84478-76-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR