2-Amino-6-chlorpurin

Lieferant: FLUOROCHEM

Synonyme: 6-Chlor-7H-purin-2-ylamin

010750-1G 010750-5G 010750-25G
FLUO010750-1GEA 0 EUR
FLUO010750-1G FLUO010750-5G FLUO010750-25G
2-Amino-6-chlorpurin
2-Amino-6-chlorpurin
Formel: C₅H₄ClN₅
Molecular Weight: 169.57 g/mol
Schmelzpunkt: 300
MDL: MFCD00075252
CAS-Nummer: 10310-21-1
EINECS: 233-686-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR