3-(Ethoxycarbonyl)phenylboronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: Ethyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 3-Ethoxycarbonylphenylboronic acid, pinacol ester, Ethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

010795-5G 010795-1G
FLUO010795-5GEA 97.4 EUR
FLUO010795-5G FLUO010795-1G
3-(Ethoxycarbonyl)phenylboronsäurepinakolester
3-(Ethoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₅H₂₁BO₄
Molecular Weight: 276.136 g/mol
MDL: MFCD03093896
CAS-Nummer: 269410-00-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR