Phenylboronsäurepinakolester
Lieferant: FLUOROCHEM
Synonyme:
(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, Benzeneboronic acid, pinacol ester, 4,4,5,5-Tetramethyl-2-phenyl-1,3,2-dioxaborolane, 2-Phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
010827-25G
FLUO010827-25GEA
107
EUR
FLUO010827-25G
Phenylboronsäurepinakolester
Phenylboronsäurepinakolester
Formel:
C₁₂H₁₇BO₂ Molecular Weight: 204,08 g/mol |
MDL:
MFCD00966985 CAS-Nummer: 24388-23-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...