2-Formylphenylboronsäure
Lieferant: FLUOROCHEM
Synonyme:
2-Boronobenzaldehyd, (2-Formylphenyl)metaborsäure, Dihydroxy(2-formylphenyl)boran, 2-Formylbenzolboronsäure
010998-5G
010998-25G
FLUO010998-5GEA
42.9
EUR
FLUO010998-5G
FLUO010998-25G
2-Formylphenylboronsäure
2-Formylphenylboronsäure
Formel:
C₇H₇BO₃ Molecular Weight: 149.94 g/mol |
MDL:
MFCD00151822 CAS-Nummer: 40138-16-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...