5-Methyl-2-thiophenboronsäure

Lieferant: FLUOROCHEM

011005-1G 011005-5G
FLUO011005-1GEA 36.9 EUR
FLUO011005-1G FLUO011005-5G
5-Methyl-2-thiophenboronsäure
5-Methyl-2-thiopheneboronic acid
Formel: C₅H₇BO₂S
MDL: MFCD01318166
CAS-Nummer: 162607-20-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR