2-(Trifluormethyl)phenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: Dihydroxy(2-(trifluormethyl)phenyl)boran, 2-(Trifluormethyl)benzolboronsäure, (2-(Trifluormethyl)phenyl)metaborsäure

011012-1G 011012-5G
FLUO011012-1GEA 0 EUR
FLUO011012-1G FLUO011012-5G
2-(Trifluormethyl)phenylboronsäure
2-(Trifluormethyl)phenylboronsäure
Formel: C₇H₆BF₃O₂
Molecular Weight: 189.93 g/mol
MDL: MFCD00236059
CAS-Nummer: 1423-27-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR