4-(Brommethyl)phenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 4-(Brommethyl)benzolboronsäure, (4-(Brommethyl)phenyl)metaborsäure, Dihydroxy(4-(brommethyl)phenyl)boran

011020-1G 011020-5G
FLUO011020-1GEA 42.9 EUR
FLUO011020-1G FLUO011020-5G
4-(Brommethyl)phenylboronsäure
4-(Brommethyl)phenylboronsäure
Formel: C₇H₈BBrO₂
MDL: MFCD01318113
CAS-Nummer: 68162-47-0
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR