4-(Brommethyl)phenylboronsäure
Lieferant: FLUOROCHEM
Synonyme:
4-(Brommethyl)benzolboronsäure, (4-(Brommethyl)phenyl)metaborsäure, Dihydroxy(4-(brommethyl)phenyl)boran
011020-1G
011020-5G
FLUO011020-1GEA
42.9
EUR
FLUO011020-1G
FLUO011020-5G
4-(Brommethyl)phenylboronsäure
4-(Brommethyl)phenylboronsäure
Formel:
C₇H₈BBrO₂ |
MDL:
MFCD01318113 CAS-Nummer: 68162-47-0 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...