4-Iodphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 4-Jodbenzolboronsäure, 4-Iodbenzolboronsäure, (4-Iodphenyl)metaborsäure, (4-Jodphenyl)metaborsäure, Dihydroxy(4-jodphenyl)boran, Dihydroxy(4-iodphenyl)boran

011042-5G
FLUO011042-5GEA 87.9 EUR
FLUO011042-5G
4-Iodphenylboronsäure
4-Iodphenylboronsäure
Formel: C₆H₆BIO₂
Molecular Weight: 247.83 g/mol
MDL: MFCD01319014
CAS-Nummer: 5122-99-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR