2-Formyl-5-methoxyphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 5-Methoxy-2-formylphenylboronic acid

011047-5G 011047-1G
FLUO011047-5GEA 495 EUR
FLUO011047-5G FLUO011047-1G
2-Formyl-5-methoxyphenylboronsäure
2-Formyl-5-methoxyphenylboronsäure
Formel: C₈H₉BO₄
Molecular Weight: 179.9657 g/mol
MDL: MFCD03001335
CAS-Nummer: 40138-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR