5-Methylfuran-2-boronsäure
Lieferant: FLUOROCHEM
Synonyme:
5-Methyl-2-furanboronic acid, 5-Methyl-2-furylboronic acid, (5-Methylfuran-2-yl)boronic acid
011049-5G
011049-1G
FLUO011049-5GEA
324
EUR
FLUO011049-5G
FLUO011049-1G
5-Methylfuran-2-boronsäure
5-Methylfuran-2-boronsäure
Formel:
C₅H₇BO₃ Molecular Weight: 125.9183 g/mol |
MDL:
MFCD01114644 CAS-Nummer: 62306-79-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...