2'-(4-Acetophenoxy)-3'-chlor-5'-(trifluormethyl)pyridin
Lieferant: FLUOROCHEM
Synonyme:
1-(4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}phenyl)ethan-1-one
011068-5G
FLUO011068-5GEA
194
EUR
FLUO011068-5G
2'-(4-Acetophenoxy)-3'-chlor-5'-(trifluormethyl)pyridin
2'-(4-Acetophenoxy)-3'-chlor-5'-(trifluormethyl)pyridin
Formel:
C₁₄H₉CIF₃NO₂ |
MDL:
MFCD00170137 CAS-Nummer: 217186-15-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...