2-Amino-4-(4-bromphenyl)thiazol
Lieferant: FLUOROCHEM
011076-25G
011076-5G
FLUO011076-25GEA
195
EUR
FLUO011076-25G
FLUO011076-5G
2-Amino-4-(4-bromphenyl)thiazol
2-Amino-4-(4-bromophenyl)thiazole
Formel:
C₉H₇BrN₂S Molecular Weight: 255.1343 g/mol |
MDL:
MFCD00051953 CAS-Nummer: 2103-94-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...