2-Brom-2-methylpropiophenon

Lieferant: FLUOROCHEM

011099-25G
FLUO011099-25GEA 0 EUR
FLUO011099-25G
2-Brom-2-methylpropiophenon
2-Brom-2-methylpropiophenon
Formel: C₁₀H₁₁BrO
MDL: MFCD00000124
CAS-Nummer: 10409-54-8
EINECS: 233-879-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR