4-Chlor-3-nitrozimtsäure

Lieferant: FLUOROCHEM

Synonyme: 3-(4-Chloro-3-nitrophenyl)prop-2-enoic acid, 3-(4-Chloro-3-nitrophenyl)acrylic acid

011119-25G
FLUO011119-25GEA 248 EUR
FLUO011119-25G
4-Chlor-3-nitrozimtsäure
4-Chlor-3-nitrozimtsäure
Formel: C₉H₆ClNO₄
Molecular Weight: 227,601 g/mol
MDL: MFCD00063311
CAS-Nummer: 20797-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR