4-Chlorchalkon

Lieferant: FLUOROCHEM

Synonyme: 4-Chlorbenzylidenacetophenon

011128-100G 011128-25G
FLUO011128-100GEA 233 EUR
FLUO011128-100G FLUO011128-25G
4-Chlorchalkon
4-Chlorchalkon
Formel: C₁₅H₁₁ClO
Molecular Weight: 242.7 g/mol
MDL: MFCD00016345
CAS-Nummer: 956-04-7
EINECS: 213-476-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR