4,7-Dichlorisatin

Lieferant: FLUOROCHEM
Achtung

011141-10G
FLUO011141-10GEA 224 EUR
FLUO011141-10G
4,7-Dichlorisatin
4,7-Dichlorisatin
Formel: C₈H₃Cl₂NO₂
MDL: MFCD00047214
CAS-Nummer: 18711-13-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR