(2,4-Dichlorphenoxy)essigsäurehydrazid
Lieferant: FLUOROCHEM
Synonyme:
(2,4-Dichlorphenoxy)acetohydrazid, (2,4-Dichlorphenoxy)acethydrazid
011144-100G
FLUO011144-100GEA
290
EUR
FLUO011144-100G
(2,4-Dichlorphenoxy)essigsäurehydrazid
(2,4-Dichlorphenoxy)essigsäurehydrazid
Formel:
(C₆H₃Cl₂)OCH₂COONHNH₂ Molecular Weight: 235,07 g/mol |
MDL:
MFCD00051972 CAS-Nummer: 28236-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...