2,3-Dimethoxyzimtsäure

Lieferant: FLUOROCHEM

Synonyme: 2,3-Dimethoxycinnamsäure, 3-(2,3-Dimethoxyphenyl)propensäure

011155-100G
FLUO011155-100GEA 368 EUR
FLUO011155-100G
2,3-Dimethoxyzimtsäure
2,3-Dimethoxyzimtsäure
Formel: C₁₁H₁₂O₄
Molecular Weight: 208,21 g/mol
CAS-Nummer: 7461-60-1
EINECS: 231-249-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR