Methyl-6-chlornicotinat

Lieferant: FLUOROCHEM

Synonyme: Methyl 6-chloronicotinate

011192-10G
FLUO011192-10GEA 0 EUR
FLUO011192-10G
Methyl-6-chlornicotinat
Methyl-6-chlornicotinat
Formel: C₇H₆ClNO₂
Molecular Weight: 171,58 g/mol
MDL: MFCD00023420
CAS-Nummer: 73781-91-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR