ω-Brom-3-nitroacetophenon

Lieferant: FLUOROCHEM

Synonyme: 2-Brom-3'-nitroacetophenon, Brommethyl-m-nitrophenylketon, 3-Nitrophenacylbromid, α-Brom-3-nitroacetophenon

011206-50G
FLUO011206-50GEA 269 EUR
FLUO011206-50G
ω-Brom-3-nitroacetophenon
ω-Brom-3-nitroacetophenon
Formel: C₈H₆BrNO₃
Molecular Weight: 244.04 g/mol
Schmelzpunkt: 90…94 °C
MDL: MFCD00024512
CAS-Nummer: 2227-64-7
EINECS: 218-764-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR