5-Phenyl-1,3-cyclohexandion

Lieferant: FLUOROCHEM

Synonyme: 5-Phenylcyclohexane-1,3-dione

011214-25G
FLUO011214-25GEA 200 EUR
FLUO011214-25G
5-Phenyl-1,3-cyclohexandion
5-Phenyl-1,3-cyclohexanedione
Formel: C₁₂H₁₂O₂
Molecular Weight: 188,23 g/mol
MDL: MFCD00051846
CAS-Nummer: 493-72-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR