5-Propyl-1,3-cyclohexandion
Lieferant: FLUOROCHEM
011216-10G
FLUO011216-10GEA
219
EUR
FLUO011216-10G
5-Propyl-1,3-cyclohexandion
5-Propyl-1,3-cyclohexanedione
Formel:
C₉H₁₄O₂ |
MDL:
MFCD00129456 CAS-Nummer: 57641-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...