2-(2-Thienyliden)-4'-phenoxyacetophenon

Lieferant: FLUOROCHEM

011225-25G
FLUO011225-25GEA 0 EUR
FLUO011225-25G
2-(2-Thienyliden)-4'-phenoxyacetophenon
2-(2-Thienyliden)-4'-phenoxyacetophenon
Formel: C₁₉H₁₄O₂S
MDL: MFCD00170623
CAS-Nummer: 333772-32-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR