2-Brom-4'-(trifluormethyl)acetophenon

Lieferant: FLUOROCHEM

Synonyme: 4-(Trifluoromethyl)phenacyl bromide, 2-Bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one

011236-1G 011236-5G 011236-25G
FLUO011236-1GEA 31.1 EUR
FLUO011236-1G FLUO011236-5G FLUO011236-25G
2-Brom-4'-(trifluormethyl)acetophenon
2-Brom-4'-(trifluormethyl)acetophenon
Formel: C₉H₆BrF₃O
Molecular Weight: 267.0400 g/mol
MDL: MFCD00126489
CAS-Nummer: 383-53-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR