4-(4-(Trifluormethyl)phenoxy)anilin
Lieferant: FLUOROCHEM
011239-1G
011239-5G
FLUO011239-1GEA
0
EUR
FLUO011239-1G
FLUO011239-5G
4-(4-(Trifluormethyl)phenoxy)anilin
4-(4-(Trifluormethyl)phenoxy)anilin
Formel:
C₁₃H₁₀F₃NO Molecular Weight: 253.220 g/mol |
MDL:
MFCD00170930 CAS-Nummer: 57478-19-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...