3,4,5-Trimethoxybenzhydrazid
Lieferant: FLUOROCHEM
Synonyme:
3,4,5-Trimethoxybenzol-1-carbohydrazid, 3,4,5-Trimethoxybenzoesäurehydrazid, 3,4,5-Trimethoxybenzohydrazid
011241-50G
FLUO011241-50GEA
281
EUR
FLUO011241-50G
3,4,5-Trimethoxybenzhydrazid
3,4,5-Trimethoxybenzhydrazid
Formel:
C₁₀H₁₄N₂O₄ Molecular Weight: 226,23 g/mol |
MDL:
MFCD00014758 CAS-Nummer: 3291-03-0 EINECS: 221-954-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...