2-Amino-4'-nitroacetophenon Hydronitroid

Lieferant: FLUOROCHEM

011305-5G
FLUO011305-5GEA 1130 EUR
FLUO011305-5G
2-Amino-4'-nitroacetophenon Hydronitroid
2-Amino-4'-nitroacetophenon Hydronitroid
Formel: C₈H₉ClN₂O₃
MDL: MFCD00025591
CAS-Nummer: 5425-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR