(S)-(+)-3-Amino-1-Boc-piperidin

Lieferant: FLUOROCHEM

Synonyme: (S)-1-Boc-3-aminopiperidine, (S)-tert-Butyl 3-aminopiperidine-1-carboxylate, (S)-(+)-3-Amino-1-BOC-piperidine

011309-1G 011309-5G
FLUO011309-1GEA 107 EUR
FLUO011309-1G FLUO011309-5G
(S)-(+)-3-Amino-1-Boc-piperidin
(S)-(+)-3-Amino-1-Boc-piperidin
Formel: C₁₀H₂₀N₂O₂
MDL: MFCD03094718
CAS-Nummer: 625471-18-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR