3-Amino-1-Boc-piperidin

Lieferant: FLUOROCHEM

Synonyme: (±)-3-Amino-1-Boc-piperidin

011310-1G 011310-5G
FLUO011310-1GEA 0 EUR
FLUO011310-1G FLUO011310-5G
3-Amino-1-Boc-piperidin
3-Amino-1-Boc-piperidin
Formel: C₁₀H₂₀N₂O₂
MDL: MFCD01861219
CAS-Nummer: 184637-48-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR