(R)-(-)-3-Amino-1-Boc-piperidin

Lieferant: FLUOROCHEM

Synonyme: tert-butyl (3R)-3-aminopiperidine-1-carboxylate, (R)-1-Boc-3-Aminopiperidine

011311-1G
FLUO011311-1GEA 34.1 EUR
FLUO011311-1G
(R)-(-)-3-Amino-1-Boc-piperidin
(R)-(-)-3-Amino-1-Boc-piperidin
Formel: C₁₀H₂₀N₂O₂
MDL: MFCD03094717
CAS-Nummer: 188111-79-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR