(S)-(-)-1-Boc-3-aminopyrrolidin

Lieferant: FLUOROCHEM

Synonyme: (S)-1-Boc-3-Aminopyrrolidine, (S)-(-)-1-BOC-3-aminopyrrolidine, tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate

011314-1G 011314-5G
FLUO011314-1GEA 29.3 EUR
FLUO011314-1G FLUO011314-5G
(S)-(-)-1-Boc-3-aminopyrrolidin
(S)-(-)-1-Boc-3-aminopyrrolidin
Formel: C₉H₁₈N₂O₂
MDL: MFCD03419271
CAS-Nummer: 147081-44-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR