(R)-(+)-1-Boc-3-aminopyrrolidin

Lieferant: FLUOROCHEM

Synonyme: (R)-1-Boc-3-aminopyrrolidine, (3R)-(+)-1-Boc-3-aminopyrrolidine, (3R)-(+)-3-Amino-1-Boc-pyrrolidine, (R)-tert-Butyl 3-aminopyrrolidine-1-carboxylate, (3R)-(+)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine, (3R)-(+)-3-Amino-1-(tert-butoxycarbonyl)pyrrolidine

011315-1G 011315-5G
FLUO011315-1GEA 31.1 EUR
FLUO011315-1G FLUO011315-5G
(R)-(+)-1-Boc-3-aminopyrrolidin
(R)-(+)-1-Boc-3-aminopyrrolidin
Formel: C₉H₁₈N₂O₂
MDL: MFCD03419272
CAS-Nummer: 147081-49-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR