3,5-Dichlorisonicotinsäure
Lieferant: FLUOROCHEM
Synonyme:
3,5-Dichloropyridine-4-carboxylic acid
011338-1G
FLUO011338-1GEA
42.9
EUR
FLUO011338-1G
3,5-Dichlorisonicotinsäure
3,5-Dichlorisonicotinsäure
Formel:
C₆H₃Cl₂NO₂ Molecular Weight: 192 g/mol |
MDL:
MFCD01861975 CAS-Nummer: 13958-93-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...