6-Aminobenzothiazol

Lieferant: FLUOROCHEM

Synonyme: Benzothiazol-6-ylamin

011370-5G
FLUO011370-5GEA 148 EUR
FLUO011370-5G
6-Aminobenzothiazol
6-Aminobenzothiazole
Formel: C₇H₆N₂S
Molecular Weight: 150.2 g/mol
MDL: MFCD00015461
CAS-Nummer: 533-30-2
EINECS: 208-559-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR