N-Boc-N-methyl-ethylendiamin

Lieferant: FLUOROCHEM

Synonyme: N-Boc-(2-Aminoethyl)-N-methylamine, N-Methylethane-1,2-diamine, N-BOC protected

011379-5G
FLUO011379-5GEA 260 EUR
FLUO011379-5G
N-Boc-N-methyl-ethylendiamin
N-Boc-N-methyl-ethylendiamin
Formel: C₈H₁₈N₂O₂
Molecular Weight: 174,24 g/mol
MDL: MFCD01317789
CAS-Nummer: 121492-06-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR