4-Aminopiperidin

Lieferant: FLUOROCHEM

Synonyme: Piperidin-4-amine

011407-10G
FLUO011407-10GEA 0 EUR
FLUO011407-10G
4-Aminopiperidin
4-Aminopiperidin
Formel: C₅H₁₂N₂
Molecular Weight: 100,16 g/mol
MDL: MFCD02179399
CAS-Nummer: 13035-19-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR