1,4-Benzodioxan-2-carbonsäure
Lieferant: FLUOROCHEM
Synonyme:
2,3-Dihydrobenzo[b][1,4]dioxine-2-carboxylic acid
011431-10G
FLUO011431-10GEA
42.9
EUR
FLUO011431-10G
1,4-Benzodioxan-2-carbonsäure
1,4-Benzodioxan-2-carbonsäure
Formel:
C₉H₈O₄ Molecular Weight: 180,16 g/mol |
MDL:
MFCD00084888 CAS-Nummer: 3663-80-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...