2'-Methoxybiphenyl-2-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 2'-Methoxy-[1,1'-biphenyl]-2-carboxylic acid

011453-500MG
FLUO011453-500MGEA 0 EUR
FLUO011453-500MG
2'-Methoxybiphenyl-2-carbonsäure
2'-Methoxybiphenyl-2-carbonsäure
Formel: C₁₄H₁₂O₃
Molecular Weight: 228,25 g/mol
MDL: MFCD03426461
CAS-Nummer: 17296-28-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR