3'-Methoxybiphenyl-2-carbonsäure

Lieferant: FLUOROCHEM

011462-500MG
FLUO011462-500MGEA 0 EUR
FLUO011462-500MG
3'-Methoxybiphenyl-2-carbonsäure
3'-Methoxybiphenyl-2-carbonsäure
Formel: C₁₄H₁₂O₃
MDL: MFCD03426465
CAS-Nummer: 38087-96-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR