4'-Methoxybiphenyl-2-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 4'-Methoxy[1,1'-biphenyl]-2-carboxylic acid, 4'-Methoxy-[1,1'-biphenyl]-2-carboxylic acid

011466-500MG
FLUO011466-500MGEA 0 EUR
FLUO011466-500MG
4'-Methoxybiphenyl-2-carbonsäure
4'-Methoxybiphenyl-2-carbonsäure
Formel: C₁₄H₁₂O₃
Molecular Weight: 228,25 g/mol
MDL: MFCD03426469
CAS-Nummer: 18110-71-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR