3',5'-Dichlorbiphenyl-2-carbonsäure

Lieferant: FLUOROCHEM

011480-500MG
FLUO011480-500MGEA 0 EUR
FLUO011480-500MG
3',5'-Dichlorbiphenyl-2-carbonsäure
3',5'-Dichlorbiphenyl-2-carbonsäure
Formel: C₁₃H₈Cl₂O₂
CAS-Nummer: 669713-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR